Merck
  • Synthesis and antitubercular evaluation of novel dibenzo[b,d]furan and 9-methyl-9H-carbazole derived hexahydro-2H-pyrano[3,2-c]quinolines via Povarov reaction.
Synthesis and antitubercular evaluation of novel dibenzo[b,d]furan and 9-methyl-9H-carbazole derived hexahydro-2H-pyrano[3,2-c]quinolines via Povarov reaction.
European journal of medicinal chemistry (2011-07-05)
Srinivas Kantevari, Thirumal Yempala, Govardhan Surineni, Balasubramanian Sridhar, Perumal Yogeeswari, Dharmarajan Sriram
ZUSAMMENFASSUNG
A series of novel hexahydro-2H-pyrano[3,2-c]quinoline analogues derived from dibenzo[b,d]furan and 9-methyl-9H-carbazole has been synthesized in very good yields through SnCl(2)·2H(2)O catalyzed one-pot Povarov reaction (imino-Diels-Alder reaction). The imines generated in situ from dibenzo[b,d]furan-2-carbaldehyde or 9-methyl-9H-carbazole-3-carbaldehyde and aromatic amines, were reacted with 3,4-dihydro-2H-pyran in a diasteroselective manner in acetonitrile at RT. These synthesized isomeric pyranoquinoline analogues have been evaluated for their in vitro antimycobacterial activity against Mycobacterium tuberculosis H37Rv (MTB) by agar dilution method. Among the 23 compounds screened, 5-(dibenzo[b,d]furan-2-yl)-9-fluoro-3,4,4a,5,6,10b-hexahydro-2H-pyrano[3,2-c]quinoline 4f, 5-(dibenzo[b,d]furan-2-yl)-9-fluoro-3,4,4a,5,6,10b-hexahydro-2H-pyrano[3,2-c]quinoline 5f and 9-fluoro-5-(9-methyl-9H-carbazol-3-yl)-3,4,4a,5,6,10b-hexa hydro-2H-pyrano[3,2-c]quinoline 7f (MIC 3.13 μg/mL) were resulted as most active antitubercular agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Isoniazid, analytical standard, ≥99% (TLC)