Merck
MACF1 alleviates aging-related osteoporosis via HES1.
Journal of cellular and molecular medicine (2021-06-17)
Chong Yin, Ye Tian, Lifang Hu, Yang Yu, Zixiang Wu, Yan Zhang, Xue Wang, Zhiping Miao, Airong Qian
ZUSAMMENFASSUNG
Ageing-related osteoporosis is becoming an emerging threat to human health along with the ageing of human population. The decreased rate of osteogenic differentiation and bone formation is the major cause of ageing-related osteoporosis. Microtubule actin cross-linking factor 1 (MACF1) is an important cytoskeletal factor that promotes osteogenic differentiation and bone formation. However, the relationship between MACF1 expression and ageing-related osteoporosis remains unclear. This study has investigated the expression pattern of MACF1 in bone tissues of ageing-related osteoporosis patients and ageing mice. The study has further elucidated the mechanism of MACF1 promoting bone formation by inhibiting HES1 expression and activity. Moreover, the therapeutic effect of MACF1 on ageing-related osteoporosis and post-menopausal osteoporosis was evaluated through in situ injection of the MACF1 overexpression plasmid. The study supplemented the molecular mechanisms between ageing and bone formation, and provided novel targets and potential therapeutic strategy for ageing-related osteoporosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerin-2-phosphat Dinatriumsalz Hydrat, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Alizarinrot S, certified by the BSC