Merck
  • PRPF19 regulates p53-dependent cellular senescence by modulating alternative splicing of MDM4 mRNA.
PRPF19 regulates p53-dependent cellular senescence by modulating alternative splicing of MDM4 mRNA.
The Journal of biological chemistry (2021-06-19)
Kimiyoshi Yano, Ryou-U Takahashi, Bunsyo Shiotani, Junko Abe, Tomoki Shidooka, Yuki Sudo, Yusuke Yamamoto, Shisei Kan, Hiroki Sakagami, Hidetoshi Tahara
ZUSAMMENFASSUNG
Alteration of RNA splicing is a hallmark of cellular senescence, which is associated with age-related disease and cancer development. However, the roles of splicing factors in cellular senescence are not fully understood. In this study, we identified the splicing factor PRPF19 as a critical regulator of cellular senescence in normal human diploid fibroblasts. PRPF19 was downregulated during replicative senescence, and PRPF19 knockdown prematurely induced senescence-like cell cycle arrest through the p53-p21 pathway. RNA-sequencing analysis revealed that PRPF19 knockdown caused a switch of the MDM4 splicing isoform from stable full-length MDM4-FL to unstable MDM4-S lacking exon 6. We also found that PRPF19 regulates MDM4 splicing by promoting the physical interaction of other splicing factors, PRPF3 and PRPF8, which are key components of the core spliceosome, U4/U6.U5 tri-snRNP. Given that MDM4 is a major negative regulator of p53, our findings imply that PRPF19 downregulation inhibits MDM4-mediated p53 inactivation, resulting in induction of cellular senescence. Thus, PRPF19 plays an important role in the induction of p53-dependent cellular senescence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Mdmx-Antikörper, Klon 8C6, clone 8C6, from mouse