Merck
LiCl induces apoptosis via CHOP/NOXA/Mcl-1 axis in human choroidal melanoma cells.
Cancer cell international (2021-02-10)
Qiuqiu Zhang, Qianwei Zhang, Huiyuan Li, Xiaofei Zhao, Han Zhang
ZUSAMMENFASSUNG
Choroidal melanoma is the most common primary intraocular malignancy that occurs in adults. Lithium Chloride Promotes Apoptosis in Human Leukemia NB4 Cells by Inhibiting Glycogen Synthase Kinase-3 Beta. In this study, we aimed to understand whether LiCl exerts anticancer effects on choroidal melanoma cells and elucidate the underlying molecular mechanisms. Human choroidal melanoma cells were treated with LiCl, and cell survival was assessed with MTT assays. Cell reproductive viability was measured by plate colony formation assays. Cell apoptosis was evaluated using flow cytometry, and proteins were detected using western blotting. A human choroidal melanoma xenograft model was established to demonstrate the effect of LiCl on human choroidal melanoma in vivo. We found that LiCl inhibited cell survival and clonogenic potential and induced apoptosis in human choroidal melanoma cells. LiCl also reduced the proliferation of choroidal melanoma cells in vivo. Moreover, the upregulation of NOXA and downregulation of Mcl-1 were responsible for LiCl-induced apoptosis. Mcl-1 overexpression obviously impaired LiCl-induced apoptosis and cleavage of caspase8, caspase9, caspase3 and PARP. Moreover, the protein expression of endoplasmic reticulum stress markers, including IRE1α, Bip, p-eIF2α, ATF4 and CHOP, were upregulated following treatment with LiCl. When CHOP expression was knocked down and cells were treated with LiCl, the protein level of NOXA was partially increased, and Mcl-1 expression was increased, while the cleavage of caspase8, caspase9, caspase3 and PARP that was induced by the LiCl was reduced compared with the vehicle treated group. Prolonged ER stress results in the activation of the apoptotic pathway. In summary, LiCl induced an endoplasmic reticulum stress response while activating intrinsic apoptosis. Furthermore, the CHOP/NOXA/Mcl-1 axis contributed to LiCl-induced apoptosis both in vitro and in vivo. The present study provides important mechanistic insight into potential cancer treatments involving LiCl and enhances the understanding of human choroidal melanoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Noxa Mouse mAb (114C307), liquid, clone 114C307, Calbiochem®