Merck
  • The ciliary protein IFT88 controls post-natal cartilage thickness and influences development of osteoarthritis.
The ciliary protein IFT88 controls post-natal cartilage thickness and influences development of osteoarthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2021-06-10)
Clarissa R Coveney, Linyi Zhu, Jadwiga Miotla-Zarebska, Bryony Stott, Ida Parisi, Vicky Batchelor, Claudia Duarte, Emer Chang, Eleanor McSorley, Tonia L Vincent, Angus Kt Wann
ZUSAMMENFASSUNG
Mechanical and biological cues drive cellular signalling in cartilage development, health, and disease. Proteins of the primary cilium, implicated in transduction of biophysiochemical signals, control cartilage formation during skeletal development, but their influence in post-natal cartilage remains unknown. Ift88fl/fl  and AggrecanCreERT2  mice were crossed to create a cartilage-specific, inducible knockout mouse AggrecanCreERT2 ;Ift88fl/fl . Tibial articular cartilage (AC) thickness was assessed, through adolescence and adulthood, by histomorphometry and integrity by OARSI score. In situ mechanisms were investigated by immunohistochemistry (IHC), RNA scope and qPCR of micro-dissected cartilage. OA was induced by surgical destabilisation (DMM). Mice voluntarily exercised using wheels. Deletion of IFT88 resulted in progressive reductions in medial AC thickness during adolescence, and marked atrophy in adulthood. At 34 weeks of age, medial thickness was reduced from 104.00μm, [100.30-110.50, 95% CI] in Ift88fl/fl to 89.42μm [84.00-93.49, 95% CI] in AggrecanCreERT2 ;Ift88fl/fl (p<0.0001), associated with reductions in calcified cartilage. Occasionally, atrophy was associated with complete, spontaneous, medial cartilage degradation. Following DMM, AggrecanCreERT2 ;Ift88fl/fl mice had increased OA scores. Atrophy in mature AC was not associated with obvious increases in aggrecanase-mediated destruction or chondrocyte hypertrophy. Of 44 candidate genes analysed, only Tcf7l2 correlated with Ift88 expression in micro-dissected cartilage. However, RNA scope revealed increased hedgehog (Hh) signalling (Gli1), associated with reductions in Ift88, in AggrecanCreERT2 ;Ift88fl/fl cartilage. Wheel exercise restored both AC thickness and levels of Hh signalling in AggrecanCreERT2 ;Ift88fl/fl . Our results demonstrate that IFT88 is chondroprotective, regulating AC thickness, potentially by thresholding a Hh response to physiological loading that controls cartilage calcification.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%