Merck
Distinctive features of SARS-CoV-2-specific T cells predict recovery from severe COVID-19.
Cell reports (2021-07-15)
Jason Neidleman, Xiaoyu Luo, Ashley F George, Matthew McGregor, Junkai Yang, Cassandra Yun, Victoria Murray, Gurjot Gill, Warner C Greene, Joshua Vasquez, Sulggi A Lee, Eliver Ghosn, Kara L Lynch, Nadia R Roan
ZUSAMMENFASSUNG
Although T cells are likely players in severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) immunity, little is known about the phenotypic features of SARS-CoV-2-specific T cells associated with recovery from severe coronavirus disease 2019 (COVID-19). We analyze T cells from 34 individuals with COVID-19 with severity ranging from mild (outpatient) to critical, culminating in death. Relative to individuals who succumbed, individuals who recovered from severe COVID-19 harbor elevated and increasing numbers of SARS-CoV-2-specific T cells capable of homeostatic proliferation. In contrast, fatal COVID-19 cases display elevated numbers of SARS-CoV-2-specific regulatory T cells and a time-dependent escalation in activated bystander CXCR4+ T cells, as assessed by longitudinal sampling. Together with the demonstration of increased proportions of inflammatory CXCR4+ T cells in the lungs of individuals with severe COVID-19, these results support a model where lung-homing T cells activated through bystander effects contribute to immunopathology, whereas a robust, non-suppressive SARS-CoV-2-specific T cell response limits pathogenesis and promotes recovery from severe COVID-19.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ionomycin Calciumsalz aus Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline