Merck
Small Molecule Targeting of Specific BAF (mSWI/SNF) Complexes for HIV Latency Reversal.
Cell chemical biology (2018-09-11)
Christine A Marian, Mateusz Stoszko, Lili Wang, Matthew W Leighty, Elisa de Crignis, Chad A Maschinot, Jovylyn Gatchalian, Benjamin C Carter, Basudev Chowdhury, Diana C Hargreaves, Jeremy R Duvall, Gerald R Crabtree, Tokameh Mahmoudi, Emily C Dykhuizen
ZUSAMMENFASSUNG
The persistence of a pool of latently HIV-1-infected cells despite combination anti-retroviral therapy treatment is the major roadblock for a cure. The BAF (mammalian SWI/SNF) chromatin remodeling complex is involved in establishing and maintaining viral latency, making it an attractive drug target for HIV-1 latency reversal. Here we report a high-throughput screen for inhibitors of BAF-mediated transcription in cells and the subsequent identification of a 12-membered macrolactam. This compound binds ARID1A-specific BAF complexes, prevents nucleosomal positioning, and relieves transcriptional repression of HIV-1. Through this mechanism, these compounds are able to reverse HIV-1 latency in an in vitro T cell line, an ex vivo primary cell model of HIV-1 latency, and in patient CD4+ T cells without toxicity or T cell activation. These macrolactams represent a class of latency reversal agents with unique mechanism of action, and can be combined with other latency reversal agents to improve reservoir targeting.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ionomycin aus Streptomyces conglobatus, ≥98% (HPLC)