Merck
Isolation and characterization of human breast cancer cells with SOX2 promoter activity.
Biochemical and biophysical research communications (2013-06-26)
Shanshan Liang, Masako Furuhashi, Rie Nakane, Seitaro Nakazawa, Houman Goudarzi, Jun-ichi Hamada, Hisashi Iizasa
ZUSAMMENFASSUNG
Sex determining region Y-box 2 (SOX2) is well known as one of the "stemness" factors and is often expressed in cancers including breast cancer. In this study, we developed a reporter system using fluorescent protein driven by the promoter for SOX2 gene to detect and isolate living SOX2-positive cells. Using this system, we determined that SOX2 promoter activities were well correlated with SOX2 mRNA expression levels in 5 breast cancer cell lines, and that the cell population with positive SOX2 promoter activity (pSp-T(+)) isolated from one of the 5 cell lines, MCF-7 cells, showed a high SOX2 protein expression and high sphere-forming activity compared with very low promoter activity (pSp-T(low/-)). The pSp-T(+) population expressed higher mRNA levels of several stemness-related genes such as CD44, ABCB1, NANOG and TWIST1 than the pSp-T(low/-) population whereas the two populations expressed CD24 at similar levels. These results suggest that the cell population with SOX2 promoter activity contains cancer stem cell (CSC)-like cells which show different expression profiles from those of CSC-marker genes previously recognized in human breast cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®