Merck
PIK3CA mutations as prognostic factor in squamous cell lung carcinoma.
Lung cancer (Amsterdam, Netherlands) (2016-12-28)
Marc McGowan, Aleksandra Silye Hoven, Marius Lund-Iversen, Steinar Solberg, Åslaug Helland, Fred R Hirsch, Odd Terje Brustugun
ZUSAMMENFASSUNG
Mutation in the PIK3CA gene is reported frequent in squamous cell carcinomas of the lung, but its potential prognostic role is still obscure. We have studied the prognostic importance of PIK3CA mutations as well as the relation to other markers in a large number of early stage lung cancers of squamous carcinoma subtype. Tumour tissue was obtained from 308 consecutively operated lung cancer patients with squamous cell carcinoma in the period 2003-2013. DNA was isolated according to standard procedures, and mutation analysis was done with either the SnapShot method and/or using PIK3CA specific primers in the Cobas system. PD-L1-expression was analysed with immunohistochemistry After thorough follow-up (median 67.6 months), overall survival and time to relapse was calculated. Tumour tissue from 102 females and 206 males were analysed. 167 (54.2%) were in stage I, 96 (31.2%) in stage II and 45 (14.6%) in stage III. PIK3CA mutation was found in 35 (11.4%) patients, most frequently in exon 20. There were no differences in sex, stage or smoking behaviour between mutated and non-mutated cases. Patients with PIK3CA mutations had a significantly longer overall survival (p=0.042) and time to relapse (p=0.030) than non-mutated cases, and the difference in time to relapse was also retained in stage I-cases (p=0.044). PD-L1-expression was less frequent among mutated cases. Our results indicate that PIK3CA mutations may confer a survival advantage in early stage squamous cell lung cancers, but further work is needed to confirm this finding.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA-Lösung, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Resazurin, powder, BioReagent
Roche
cOmplete-ULTRA-Mini-Tabletten, Protease-Inhibitor-Cocktail in EASYpacks , Tablets supplied in foil blister packs.
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse