Merck
Novel bioassay using Bacillus megaterium to detect tetracycline in milk.
Revista Argentina de microbiologia (2016-05-02)
Melisa Tumini, Orlando G Nagel, Pilar Molina, Rafael L Althaus
ZUSAMMENFASSUNG
Tetracyclines are used for the prevention and control of dairy cattle diseases. Residues of these drugs can be excreted into milk. Thus, the aim of this study was to develop a microbiological method using Bacillus megaterium to detect tetracyclines (chlortetracycline, oxytetracycline and tetracycline) in milk. In order to approximate the limits of detection of the bioassay to the Maximum Residue Limit (100μg/l) for milk tetracycline, different concentrations of chloramphenicol (0, 1000, 1500 and 2000μg/l) were tested. The detection limits calculated were similar to the Maximum Residue Limits when a bioassay using B. megaterium ATCC 9885 spores (2.8×10(8)spores/ml) and chloramphenicol (2000μg/l) was utilized. This bioassay detects 105μg/l of chlortetracycline, 100μg/l of oxytetracycline and 134μg/l of tetracycline in 5h. Therefore, this method is suitable to be incorporated into a microbiological multi-residue system for the identification of tetracyclines in milk.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
Toluidinblau, suitable for microscopy, 8.74% (ZN (THEORY))