Merck
  • An efficient synthesis of highly functionalized novel chromeno[4,3-b]pyrroles and indolizino[6,7-b]indoles as potent antimicrobial and antioxidant agents.
An efficient synthesis of highly functionalized novel chromeno[4,3-b]pyrroles and indolizino[6,7-b]indoles as potent antimicrobial and antioxidant agents.
Bioorganic & medicinal chemistry letters (2012-01-11)
Natarajan Arumugam, Raghavachary Raghunathan, Abdulrahman I Almansour, Usama Karama
ZUSAMMENFASSUNG
A facile and efficient synthesis of novel chromeno[4,3-b]pyrroles has been accomplished by intramolecular 1,3-dipolar cycloaddition which on subsequent Pictet-Spengler cyclisation in presence of p-toluenesulfonic acid yielded indolizino[6,7-b]indoles. The synthesized chromenopyrroles and indolizinoindoles were evaluated for their antimicrobial and antioxidant activities. Compounds 7b, 7e, 7a and 7d exhibited respectively, good antibacterial and antifungal activities against tested pathogens when compared to reference control.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Carbendazim, 97%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
Carbendazim, PESTANAL®, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.