- Structure-based design, synthesis and structure-activity relationships of dibenzosuberyl- and benzoate-substituted tropines as ligands for acetylcholine-binding protein.
Structure-based design, synthesis and structure-activity relationships of dibenzosuberyl- and benzoate-substituted tropines as ligands for acetylcholine-binding protein.
Bioorganic & medicinal chemistry letters (2012-01-17)
Ewald Edink, Atilla Akdemir, Chimed Jansen, René van Elk, Obbe Zuiderveld, Frans J J de Kanter, Jacqueline E van Muijlwijk-Koezen, August B Smit, Rob Leurs, Iwan J P de Esch
PMID22243960
ZUSAMMENFASSUNG
Using structure-based optimization procedures on in silico hits, dibenzosuberyl- and benzoate substituted tropines were designed as ligands for acetylcholine-binding protein (AChBP). This protein is a homolog to the ligand binding domain of the nicotinic acetylcholine receptor (nAChR). Distinct SAR is observed between two AChBP species variants and between the α7 and α4β2 nAChR subtype. The AChBP species differences are indicative of a difference in accessibility of a ligand-inducible subpocket. Hereby, we have identified a region that can be scrutinized to achieve selectivity for nicotinic receptor subtypes.