- Praziquantel derivatives exhibit activity against both juvenile and adult Schistosoma japonicum.
Praziquantel derivatives exhibit activity against both juvenile and adult Schistosoma japonicum.
Bioorganic & medicinal chemistry letters (2012-01-24)
Wen-wen Duan, Si-jie Qiu, Yue Zhao, Huan Sun, Chunhua Qiao, Chao-ming Xia
PMID22264473
ZUSAMMENFASSUNG
A praziquantel analog 10-hydroxy praziquantel and eight praziquantel/peroxide conjugates were synthesized. The biological activity of these compounds was evaluated against juvenile and adult stages of Schistosoma japonicum. Unlike praziquantel, 10-hydroxy praziquantel exhibits activity against both juvenile and adult Schistosoma japonicumin. All hybrid compounds displayed modest to significant worm killing activity. The present study has important significance for the development of hybrid antischistosomal drugs.