Merck
Structural basis for recruitment of the CHK1 DNA damage kinase by the CLASPIN scaffold protein.
Structure (London, England : 1993) (2021-04-01)
Matthew Day, Sarah Parry-Morris, Jack Houghton-Gisby, Antony W Oliver, Laurence H Pearl
ZUSAMMENFASSUNG
CHK1 is a protein kinase that functions downstream of activated ATR to phosphorylate multiple targets as part of intra-S and G2/M DNA damage checkpoints. Its role in allowing cells to survive replicative stress has made it an important target for anti-cancer drug discovery. Activation of CHK1 by ATR depends on their mutual interaction with CLASPIN, a natively unstructured protein that interacts with CHK1 through a cluster of phosphorylation sites in its C-terminal half. We have now determined the crystal structure of the kinase domain of CHK1 bound to a high-affinity motif from CLASPIN. Our data show that CLASPIN engages a conserved site on CHK1 adjacent to the substrate-binding cleft, involved in phosphate sensing in other kinases. The CLASPIN motif is not phosphorylated by CHK1, nor does it affect phosphorylation of a CDC25 substrate peptide, suggesting that it functions purely as a scaffold for CHK1 activation by ATR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
cOmplete, EDTA-freier Proteasehemmer-Cocktail, Tablets provided in glass vials
Sigma-Aldrich
Adenosin-5′-[γ-thio]triphosphat Tetralithiumsalz, ≥75% (HPLC), powder