Merck
  • Properdin binds independent of complement activation in an in vivo model of anti-glomerular basement membrane disease.
Properdin binds independent of complement activation in an in vivo model of anti-glomerular basement membrane disease.
Kidney international (2018-10-17)
Joseph O'Flynn, Juha Kotimaa, Ria Faber-Krol, Karin Koekkoek, Ngaisah Klar-Mohamad, Angela Koudijs, Wilhelm J Schwaeble, Cordula Stover, Mohamed R Daha, Cees van Kooten
ZUSAMMENFASSUNG
Properdin is the only known positive regulator of complement activation by stabilizing the alternative pathway convertase through C3 binding, thus prolonging its half-life. Recent in vitro studies suggest that properdin may act as a specific pattern recognition molecule. To better understand the role of properdin in vivo, we used an experimental model of acute anti-glomerular basement membrane disease with wild-type, C3- and properdin knockout mice. The model exhibited severe proteinuria, acute neutrophil infiltration and activation, classical and alternative pathway activation, and progressive glomerular deposition of properdin, C3 and C9. Although the acute renal injury was likely due to acute neutrophil activation, we found properdin deposition in C3-knockout mice that was not associated with IgG. Thus, properdin may deposit in injured tissues in vivo independent of its main ligand C3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Anti-Digoxigenin-POD, Fab-Fragmente, from sheep