Merck
Soluble Aβ oligomers and protofibrils induce NLRP3 inflammasome activation in microglia.
Journal of neurochemistry (2019-12-25)
Asta Lučiūnaitė, Róisín M McManus, Marija Jankunec, Ildikó Rácz, Cira Dansokho, Indrė Dalgėdienė, Stephanie Schwartz, Frederic Brosseron, Michael T Heneka
ZUSAMMENFASSUNG
Alzheimer's disease (AD) is the most prevalent neurodegenerative disorder causing memory loss, language problems and behavioural disturbances. AD is associated with the accumulation of fibrillar amyloid-β (Aβ) and the formation of neurofibrillary tau tangles. Fibrillar Aβ itself represents a danger-associated molecular pattern, which is recognized by specific microglial receptors. One of the key players is formation of the NOD-, LRR- and pyrin domain-containing 3 (NLRP3) inflammasome, whose activation has been demonstrated in AD patient brains and transgenic animal models of AD. Here, we investigated whether Aβ oligomers or protofibrils that represent lower molecular aggregates prior to Aβ deposition are able to activate the NLRP3 inflammasome and subsequent interleukin-1 beta (IL-1β) release by microglia. In our study, we used Aβ preparations of different sizes: small oligomers and protofibrils of which the structure was confirmed by atomic force microscopy. Primary microglial cells from C57BL/6 mice were treated with the respective Aβ preparations and NLRP3 inflammasome activation, represented by caspase-1 cleavage, IL-1β production, and apoptosis-associated speck-like protein containing a CARD speck formation was analysed. Both protofibrils and low molecular weight Aβ aggregates induced a significant increase in IL-1β release. Inflammasome activation was confirmed by apoptosis-associated speck-like protein containing a CARD speck formation and detection of active caspase-1. The NLRP3 inflammasome inhibitor MCC950 completely inhibited the Aβ-induced immune response. Our results show that the NLRP3 inflammasome is activated not only by fibrillar Aβ aggregates as reported before, but also by lower molecular weight Aβ oligomers and protofibrils, highlighting the possibility that microglial activation by these Aβ species may initiate innate immune responses in the central nervous system prior to the onset of Aβ deposition. Cover Image for this issue: https://doi.org/10.1111/jnc.14773.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Paraformaldehyd, powder, 95%
Roche
Cytotoxicity Detection Kit (LDH), suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests
Sigma-Aldrich
Azurin aus Pseudomonas aeruginosa, from Pseudomonas aeruginosa, lyophilized powder