Merck
  • Structural insights into hormone recognition by the human glucose-dependent insulinotropic polypeptide receptor.
Structural insights into hormone recognition by the human glucose-dependent insulinotropic polypeptide receptor.
eLife (2021-07-14)
Fenghui Zhao, Chao Zhang, Qingtong Zhou, Kaini Hang, Xinyu Zou, Yan Chen, Fan Wu, Qidi Rao, Antao Dai, Wanchao Yin, Dan-Dan Shen, Yan Zhang, Tian Xia, Raymond C Stevens, H Eric Xu, Dehua Yang, Lihua Zhao, Ming-Wei Wang
ZUSAMMENFASSUNG
Glucose-dependent insulinotropic polypeptide (GIP) is a peptide hormone that exerts crucial metabolic functions by binding and activating its cognate receptor, GIPR. As an important therapeutic target, GIPR has been subjected to intensive structural studies without success. Here, we report the cryo-EM structure of the human GIPR in complex with GIP and a Gs heterotrimer at a global resolution of 2.9 Å. GIP adopts a single straight helix with its N terminus dipped into the receptor transmembrane domain (TMD), while the C terminus is closely associated with the extracellular domain and extracellular loop 1. GIPR employs conserved residues in the lower half of the TMD pocket to recognize the common segments shared by GIP homologous peptides, while uses non-conserved residues in the upper half of the TMD pocket to interact with residues specific for GIP. These results provide a structural framework of hormone recognition and GIPR activation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, powder
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥3.0 units/mg protein, lyophilized powder