Merck
Epithelial response to IFN-γ promotes SARS-CoV-2 infection.
EMBO molecular medicine (2021-02-06)
Julian Heuberger, Jakob Trimpert, Daria Vladimirova, Christian Goosmann, Manqiang Lin, Rosa Schmuck, Hans-Joachim Mollenkopf, Volker Brinkmann, Frank Tacke, Nikolaus Osterrieder, Michael Sigal
ZUSAMMENFASSUNG
SARS-CoV-2, the agent that causes COVID-19, invades epithelial cells, including those of the respiratory and gastrointestinal mucosa, using angiotensin-converting enzyme-2 (ACE2) as a receptor. Subsequent inflammation can promote rapid virus clearance, but severe cases of COVID-19 are characterized by an inefficient immune response that fails to clear the infection. Using primary epithelial organoids from human colon, we explored how the central antiviral mediator IFN-γ, which is elevated in COVID-19, affects epithelial cell differentiation, ACE2 expression, and susceptibility to infection with SARS-CoV-2. In mouse and human colon, ACE2 is mainly expressed by surface enterocytes. Inducing enterocyte differentiation in organoid culture resulted in increased ACE2 production. IFN-γ treatment promoted differentiation into mature KRT20+ enterocytes expressing high levels of ACE2, increased susceptibility to SARS-CoV-2 infection, and resulted in enhanced virus production in infected cells. Similarly, infection-induced epithelial interferon signaling promoted enterocyte maturation and enhanced ACE2 expression. We here reveal a mechanism by which IFN-γ-driven inflammatory responses induce a vulnerable epithelial state with robust replication of SARS-CoV-2, which may have an impact on disease outcome and virus transmission.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
N-Acetyl-L-Cystein, suitable for cell culture, BioReagent
Sigma-Aldrich
Gentamicin -Lösung, 10 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Methylzellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
TGF-β RI Kinase-Inhibitor IV, CAS-Nr. 909910-43-6, Calbiochem, TGF-β RI Kinase Inhibitor IV, CAS 909910-43-6, is a cell-permeable, selective inhibitor of ALK-4/5/7-mediated signaling (IC50 = 45, 12, and 7.5 nM, respectively).
Sigma-Aldrich
Anti-ACE2 in Kaninchen hergestellte Antikörper, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution