Merck
  • HOXC10 promotes proliferation and invasion and induces immunosuppressive gene expression in glioma.
HOXC10 promotes proliferation and invasion and induces immunosuppressive gene expression in glioma.
The FEBS journal (2018-04-21)
Shu Li, Wenhao Zhang, Chao Wu, Hongliang Gao, Jun Yu, Xiaoqiang Wang, Bin Li, Zhong Jun, Wenchaun Zhang, Ping Zhou, Juanhong Shi, Lifeng Wang, Yunxing Gao, Shiting Li, Bangbao Tao
ZUSAMMENFASSUNG
The prognosis for patients with malignant glioma is very poor and thus the identification of new potential therapeutic targets is critically important. In this work, we report a previously unknown role for the homeobox transcription factor HOXC10 in regulating immunosuppressive gene expression in glioma cell lines and their proliferative and invasive capacities. Although HOXC10 expression is dysregulated in several types of tumors, its potential function in glioma was not known. We found that HOXC10 expression was upregulated in glioma compared with normal tissue, and that HOXC10 expression positively associated with high grading of glioma. In three independent datasets (REMBRANDT glioma, The Cancer Genome Atlas glioblastoma multiforme and GSE4412), HOXC10 upregulation was associated with short overall survival. In two glioma cell lines, HOXC10 knock-down inhibited cell proliferation, colony formation, migration and invasion, and promoted apoptosis. In addition, HOXC10 knock-down suppressed the expression of genes that are involved in tumor immunosuppression, including those for transforming growth factor-β 2, PD-L2, CCL2 and TDO2. A ChIP assay showed that HOXC10 directly bound to the PD-L2 and TDO2 promoter regions. In summary, our results suggest that HOXC10 upregulation in glioma promotes an aggressive phenotype and induces immunosuppressive gene expression, supporting further investigation of the potential of HOXC10 as a therapeutic target in glioma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-HOXC10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution