Merck
Germline transgenesis in pigs by cytoplasmic microinjection of Sleeping Beauty transposons.
Nature protocols (2014-03-15)
Zoltán Ivics, Wiebke Garrels, Lajos Mátés, Tien Yin Yau, Sanum Bashir, Vaclav Zidek, Vladimír Landa, Aron Geurts, Michal Pravenec, Thomas Rülicke, Wilfried A Kues, Zsuzsanna Izsvák
ZUSAMMENFASSUNG
The pig has emerged as an important large animal model in biomedical and pharmaceutical research. We describe a protocol for high-efficiency germline transgenesis and sustained transgene expression in pigs by using the Sleeping Beauty (SB) transposon system. The protocol is based on co-injection of a plasmid encoding the SB100X hyperactive transposase, together with a second plasmid carrying a transgene flanked by binding sites for the transposase, into the cytoplasm of porcine zygotes. The transposase mediates excision of the transgene cassette from the plasmid vector and its permanent insertion into the genome to produce stable transgenic animals. This method compares favorably in terms of both efficiency and reliable transgene expression to classic pronuclear microinjection or somatic cell nuclear transfer (SCNT), and it offers comparable efficacies to lentiviral approaches, without limitations on vector design, issues of transgene silencing and the toxicity and biosafety concerns of working with viral vectors. Microinjection of the vectors into zygotes and transfer of the embryos to recipient animals can be performed in 1 d; generation of germline-transgenic lines by using this protocol takes ∼1 year.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Natrium-L-lactat, ≥99.0% (NT)
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Tris-EDTA-Puffer -Lösung, BioUltra, Molecular Biology, pH 8.0
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Hypotaurin, ≥98% (TLC)
Sigma-Aldrich
Calciumchlorid Dihydrat, BioXtra, ≥99.0%