Merck
  • The RAS GTPase RIT1 compromises mitotic fidelity through spindle assembly checkpoint suppression.
The RAS GTPase RIT1 compromises mitotic fidelity through spindle assembly checkpoint suppression.
Current biology : CB (2021-07-09)
Antonio Cuevas-Navarro, Richard Van, Alice Cheng, Anatoly Urisman, Pau Castel, Frank McCormick
ZUSAMMENFASSUNG
The spindle assembly checkpoint (SAC) functions as a sensor of unattached kinetochores that delays mitotic progression into anaphase until proper chromosome segregation is guaranteed.1,2 Disruptions to this safety mechanism lead to genomic instability and aneuploidy, which serve as the genetic cause of embryonic demise, congenital birth defects, intellectual disability, and cancer.3,4 However, despite the understanding of the fundamental mechanisms that control the SAC, it remains unknown how signaling pathways directly interact with and regulate the mitotic checkpoint activity. In response to extracellular stimuli, a diverse network of signaling pathways involved in cell growth, survival, and differentiation are activated, and this process is prominently regulated by the Ras family of small guanosine triphosphatases (GTPases).5 Here we show that RIT1, a Ras-related GTPase that regulates cell survival and stress response,6 is essential for timely progression through mitosis and proper chromosome segregation. RIT1 dissociates from the plasma membrane (PM) during mitosis and interacts directly with SAC proteins MAD2 and p31comet in a process that is regulated by cyclin-dependent kinase 1 (CDK1) activity. Furthermore, pathogenic levels of RIT1 silence the SAC and accelerate transit through mitosis by sequestering MAD2 from the mitotic checkpoint complex (MCC). Moreover, SAC suppression by pathogenic RIT1 promotes chromosome segregation errors and aneuploidy. Our results highlight a unique function of RIT1 compared to other Ras GTPases and elucidate a direct link between a signaling pathway and the SAC through a novel regulatory mechanism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Proteasehemmer-Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Trypsin, Singles, Proteomik-Qualität
Sigma-Aldrich
Anti-Histidin-markiert-Antikörper, Klon HIS.H8, clone HIS.H8, Upstate®, from mouse
Sigma-Aldrich
Anti-p31comet-Antikörper, Klon E29.19.14, clone E29.19.14, from mouse