Merck
  • Synthesis and preliminary antibacterial evaluation of Linezolid-like 1,2,4-oxadiazole derivatives.
Synthesis and preliminary antibacterial evaluation of Linezolid-like 1,2,4-oxadiazole derivatives.
European journal of medicinal chemistry (2012-03-01)
Antonio Palumbo Piccionello, Rosario Musumeci, Clementina Cocuzza, Cosimo Gianluca Fortuna, Annalisa Guarcello, Paola Pierro, Andrea Pace
ZUSAMMENFASSUNG
In the present study the synthesis of new Linezolid-like molecules has been achieved by substitution of the oxazolidinone central heterocyclic moiety with a 1,2,4-oxadiazole ring. Two series of 1,2,4-oxadiazoles, bearing different side-chains and containing a varying number of fluorine atoms, were synthesized and preliminarily tested for biological activity against some Gram-positive and Gram-negative bacteria using Linezolid and Ceftriaxone as reference drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.