Merck
Characterization and in vitro culture of male germ cells from developing bovine testis.
The Journal of reproduction and development (2011-02-04)
Mayako Fujihara, Sung-Min Kim, Naojiro Minami, Masayasu Yamada, Hiroshi Imai
ZUSAMMENFASSUNG
The transition from male primitive germ cells (gonocytes) to type A spermatogonia in the neonatal testis is the initial process and a crucial process in spermatogenesis. However, in large domestic animals, the physiological and biochemical characteristics of germ cells during the developmental processes remain largely unknown. In this study, we characterized bovine germ cells in the developing testis from the neonatal stage to the adult stage. The binding of the lectin Dolichos biflorus agglutinin (DBA) and the expression of ubiquitin carboxyl-terminal hydrolase 1 (UCHL1) were restricted to gonocytes in the neonatal testis and spermatogonia in the adult testis. Gonocytes also expressed a germ cell marker (VASA) and stem cell markers (NANOG and OCT3/4), while the expressions of these markers in the adult testis were restricted to differentiated spermatic cells and were rarely expressed in spermatogonia. We subsequently utilized these markers to characterize gonocytes and spermatogonia after culture in vitro. Spermatogonia that were collected from the adult testis formed colonies in vitro only for one week. On the other hand, gonocytes from the neonatal testis could proliferate and form colonies after every passage for 1.5 months in culture. These colonies retained undifferentiated states of gonocytes as confirmed by the expression of both germ cell and stem cell markers. Moreover, a transplantation assay using immunodeficient mice testes showed that long-term cultured cells derived from gonocytes were able to colonize in the recipient testis. These results indicated that bovine gonocytes could maintain germ cell and stem cell potential in vitro.