- Benzotriazol-1-yl-sulfonyl azide for diazotransfer and preparation of azidoacylbenzotriazoles.
Benzotriazol-1-yl-sulfonyl azide for diazotransfer and preparation of azidoacylbenzotriazoles.
The Journal of organic chemistry (2010-09-10)
Alan R Katritzky, Mirna El Khatib, Oleg Bol'shakov, Levan Khelashvili, Peter J Steel
PMID20825153
ZUSAMMENFASSUNG
Benzotriazol-1-yl-sulfonyl azide, a new crystalline, stable, and easily available diazotransfer reagent provides N-(α-azidoacyl)benzotriazoles convenient for N-, O-, C- and S-acylations. The efficient syntheses of various amides, azido protected peptides, esters, ketones and thioesters is reported together with a wide range of azides (including α-azido acids from α- amino acids in partially aqueous conditions) and diazo compounds.