Merck
  • Increased hemoglobin and heme in MALDI-TOF MS analysis induce ferroptosis and promote degeneration of herniated human nucleus pulposus.
Increased hemoglobin and heme in MALDI-TOF MS analysis induce ferroptosis and promote degeneration of herniated human nucleus pulposus.
Molecular medicine (Cambridge, Mass.) (2021-09-10)
Liang Shan, Ximing Xu, Jing Zhang, Peng Cai, Han Gao, Yingjie Lu, Jiangang Shi, Yinlong Guo, Yue Su
ZUSAMMENFASSUNG
Neovasculogenesis is characteristic of herniated lumbar discs, in which extruded nucleus pulposus is prone to heme iron-induced cytotoxicity (increased oxidative stress causing ferroptosis). However, recent analyses of neovascularization are very complicated, and the mechanism of action is rarely reported. Matrix-assisted laser desorption/ionization-time-of-flight mass spectrometry (MALDI-TOF MS) was performed to analyze human herniated and nonherniated nucleus pulposus. Then, the clinical relevance of the MALDI-TOF MS results and Pfirrmann classification of the degenerative nucleus pulposus were analyzed. To explore the mechanism, the heme-induced ferroptosis effect was evaluated at both the tissue and cell levels using high-resolution MALDI-TOF MS and molecular biology methods. The spectra revealed that hemoglobin (Hb) and heme signals were greatly increased, thus serving as predictors of vasculogenesis in herniated nucleus pulposus. The clinical relevance analysis demonstrated that the intensity of Hb and heme peaks was closely related to the Pfirrmann classification of degenerative nucleus pulposus. Mechanistically, increased heme catabolism and downregulation of glutathione peroxidase 4 (GPX4) levels were detected in herniated nucleus pulposus, reflecting iron-dependent cell death or ferroptosis. Iron levels was also increased in herniated nucleus pulposus compared with that in nonherniated nucleus pulposus. Furthermore, accuracy mass measurements confirmed that the levels of ferroptosis-related metabolites, such as glutathione, arachidonic acid (AA), sphinganine, polyunsaturated fatty acid (PUFA), and tricarboxylic acid (TCA) cycle metabolites, were significantly different between herniated and nonherniated tissues, indicating that the interior of the herniated tissues is a pro-oxidant environment. Moreover, heme-induced ferroptosis was verified in human nucleus pulposus cells (HNPCs), and the underlying mechanism might be associated with the Notch pathway. Neovascularization in herniated nucleus pulposus may expose tissues to high levels of heme, which can induce cytotoxicity and ferroptosis within tissues and accelerate the progressive degeneration of herniated nucleus pulposus. This study is beneficial for understanding the pathological mechanism of herniated nucleus pulposus and facilitating the development of nonoperative interventions for treating lumbar disc herniation (LDH).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumeisen(III)-Citrat, reagent grade, powder
Sigma-Aldrich
Hämoglobin human, lyophilized powder
Sigma-Aldrich
Erastin, ≥98% (HPLC)