Merck
  • Isolated downregulation of HCN2 in ventricles of rats with streptozotocin-induced diabetic cardiomyopathy.
Isolated downregulation of HCN2 in ventricles of rats with streptozotocin-induced diabetic cardiomyopathy.
BMC cardiovascular disorders (2021-03-04)
Katarina Hadova, Eva Kralova, Gabriel Doka, Lenka Bies Pivackova, Zuzana Kmecova, Peter Krenek, Jan Klimas
ZUSAMMENFASSUNG
In spite of disrupted repolarization of diabetic heart, some studies report less tendency of diabetic heart to develop ventricular arrhythmias suggesting effective compensatory mechanism. We hypothesized that myocardial alterations in HCN2 and HCN4 channels occur under hyperglycaemia. Diabetes was induced in rats using a single injection of streptozotocin (STZ; 55 mg/kg body weight, i.p.). Basal ECG was measured. Expression of mRNA for HCN channels, potassium channels and microRNA 1 and 133a were measured in ventricular tissues. Protein expression of HCN2 channel isoform was assessed in five different regions of the heart by western blotting. Differentiated H9c2 cell line was used to examine HCN channels expression under hyperglycaemia in vitro. Six weeks after STZ administration, heart rate was reduced, QRS complex duration, QT interval and T-wave were prolonged in diabetic rats compared to controls. mRNA and protein expressions of HCN2 decreased exclusively in the ventricles of diabetic rats. HCN2 expression levels in atria of STZ rats and H9c2 cells treated with excess of glucose were not changed. MicroRNA levels were stable in STZ rat hearts. We found significantly decreased mRNA levels of several potassium channels participating in repolarization, namely Kcnd2 (Ito1), Kcnh2 (IKr), Kcnq1 (IKs) and Kcnj11 (IKATP). This result together with downregulated HCN2 channels suggest that HCN channels might be an integral part of ventricular electric remodelling and might play a role in cardiac repolarization projected in altered arrhythmogenic profile of diabetic heart.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-α-Aktinin (Sarkomer) in Maus hergestellte Antikörper, clone EA-53, ascites fluid