- Astrocyte Ca2+-evoked ATP release regulates myelinated axon excitability and conduction speed.
Astrocyte Ca2+-evoked ATP release regulates myelinated axon excitability and conduction speed.
Science (New York, N.Y.) (2021-10-15)
Jonathan Lezmy, I Lorena Arancibia-Cárcamo, Tania Quintela-López, Diane L Sherman, Peter J Brophy, David Attwell
PMID34648330
ZUSAMMENFASSUNG
[Figure: see text].
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2, clone 7F6-G5-A2, Upstate®, from mouse