Merck
  • N-terminal selective conjugation method widens the therapeutic window of antibody-drug conjugates by improving tolerability and stability.
N-terminal selective conjugation method widens the therapeutic window of antibody-drug conjugates by improving tolerability and stability.
mAbs (2021-04-28)
Min Ji Ko, Daehae Song, Juhee Kim, Jae Yong Kim, Jaehyun Eom, Byungje Sung, Yong-Gyu Son, Young Min Kim, Sang Hoon Lee, Weon-Kyoo You, Jinwon Jung
ZUSAMMENFASSUNG
Antibody-drug conjugates (ADCs) are targeted therapeutic agents that treat cancers by selective delivery of highly potent cytotoxic drugs to tumor cells via cancer-specific antibodies. However, their clinical benefit is limited by off-target toxicity and narrow therapeutic windows. To overcome these limitations, we have applied reductive alkylation to develop a new type of ADC that has cytotoxic drugs conjugated to the N-terminal of an antibody through amine bonds introduced via reductive alkylation reactions (NTERM). To test whether the NTERM-conjugated ADCs can widen therapeutic windows, we synthesized three different ADCs by conjugating trastuzumab and monomethyl auristatin-F using three different methods, and compared their stability, efficacy, and toxicity. The NTERM-conjugated ADC was more stable in vitro and in vivo than the thiol-conjugated and the lysine-conjugated ADCs. The NTERM-conjugated ADC showed lower toxicity compared to other ADCs, whereas its efficacy was comparable to that of the thiol-conjugated ADC and better than that of the lysine-conjugated ADC. These results suggest that the NTERM conjugation method could widen the therapeutic window of ADCs by enhancing its stability and reducing toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,3′,5,5′-3,3,5,5-Tetramethylbenzidin (TMB) Flüssigsubstratsystem für ELISA, peroxidase substrate
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution