Merck
Energetic and structural features of SARS-CoV-2 N-protein co-assemblies with nucleic acids.
bioRxiv : the preprint server for biology (2021-02-18)
Huaying Zhao, Di Wu, Ai Nguyen, Yan Li, Regina C Adão, Eugene Valkov, George H Patterson, Grzegorz Piszczek, Peter Schuck
ZUSAMMENFASSUNG
Nucleocapsid (N) protein of the SARS-CoV-2 virus packages the viral genome into well-defined ribonucleoprotein particles, but the molecular pathway is still unclear. N-protein is dimeric and consists of two folded domains with nucleic acid (NA) binding sites, surrounded by intrinsically disordered regions that promote liquid-liquid phase separation. Here we use biophysical tools to study N-protein interactions with oligonucleotides of different length, examining the size, composition, secondary structure, and energetics of the resulting states. We observe formation of supramolecular clusters or nuclei preceding growth into phase-separated droplets. Short hexanucleotide NA forms compact 2:2 N-protein/NA complexes with reduced disorder. Longer oligonucleotides expose additional N-protein interactions and multi-valent protein-NA interactions, which generate higher-order mixed oligomers and simultaneously promote growth of droplets. Phase separation is accompanied by a significant increase in protein secondary structure, different from that caused by initial NA binding, which may contribute to the assembly of ribonucleoprotein particles within molecular condensates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%