Merck
Generation and Culture of Lingual Organoids Derived from Adult Mouse Taste Stem Cells.
Journal of visualized experiments : JoVE (2021-04-20)
Lauren A Shechtman, Christina M Piarowski, Jennifer K Scott, Erin J Golden, Dany Gaillard, Linda A Barlow
ZUSAMMENFASSUNG
The sense of taste is mediated by taste buds on the tongue, which are composed of rapidly renewing taste receptor cells (TRCs). This continual turnover is powered by local progenitor cells and renders taste function prone to disruption by a multitude of medical treatments, which in turn severely impacts the quality of life. Thus, studying this process in the context of drug treatment is vital to understanding if and how taste progenitor function and TRC production are affected. Given the ethical concerns and limited availability of human taste tissue, mouse models, which have a taste system similar to humans, are commonly used. Compared to in vivo methods, which are time-consuming, expensive, and not amenable to high throughput studies, murine lingual organoids can enable experiments to be run rapidly with many replicates and fewer mice. Here, previously published protocols have been adapted and a standardized method for generating taste organoids from taste progenitor cells isolated from the circumvallate papilla (CVP) of adult mice is presented. Taste progenitor cells in the CVP express LGR5 and can be isolated via EGFP fluorescence-activated cell sorting (FACS) from mice carrying an Lgr5EGFP-IRES-CreERT2 allele. Sorted cells are plated onto a matrix gel-based 3D culture system and cultured for 12 days. Organoids expand for the first 6 days of the culture period via proliferation and then enter a differentiation phase, during which they generate all three taste cell types along with non-taste epithelial cells. Organoids can be harvested upon maturation at day 12 or at any time during the growth process for RNA expression and immunohistochemical analysis. Standardizing culture methods for production of lingual organoids from adult stem cells will improve reproducibility and advance lingual organoids as a powerful drug screening tool in the fight to help patients experiencing taste dysfunction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Roche
Dispase® II (neutrale Protease, Klasse II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Paraformaldehyd, powder, 95%