Merck
A Specialized Niche in the Pancreatic Microenvironment Promotes Endocrine Differentiation.
Developmental cell (2020-08-29)
Corinna Cozzitorto, Laura Mueller, Silvia Ruzittu, Nancy Mah, David Willnow, Jean-Francois Darrigrand, Heather Wilson, Daniel Khosravinia, Amir-Ala Mahmoud, Maurizio Risolino, Licia Selleri, Francesca M Spagnoli
ZUSAMMENFASSUNG
The interplay between pancreatic epithelium and the surrounding microenvironment is pivotal for pancreas formation and differentiation as well as adult organ homeostasis. The mesenchyme is the main component of the embryonic pancreatic microenvironment, yet its cellular identity is broadly defined, and whether it comprises functionally distinct cell subsets is not known. Using genetic lineage tracing, transcriptome, and functional studies, we identified mesenchymal populations with different roles during pancreatic development. Moreover, we showed that Pbx transcription factors act within the mouse pancreatic mesenchyme to define a pro-endocrine specialized niche. Pbx directs differentiation of endocrine progenitors into insulin- and glucagon-positive cells through non-cell-autonomous regulation of ECM-integrin interactions and soluble molecules. Next, we measured functional conservation between mouse and human pancreatic mesenchyme by testing identified mesenchymal factors in an iPSC-based differentiation model. Our findings provide insights into how lineage-specific crosstalk between epithelium and neighboring mesenchymal cells underpin the generation of different pancreatic cell types.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ROCK-Inhibitor (Y-27632), The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Anti-α-Amylase antibody produced in rabbit, fractionated antiserum, lyophilized powder
Sigma-Aldrich
αα-Amyloidpräkursorprotein-Modulator, A cell-permeable benzolactam derived PKC activator (Ki = 11.9 nM for PKCα) that efficiently enhances non-amyloidogenic α-processing of amyloid precursor protein (APP) even at 100 nM in human fibroblast AG06848.
Sigma-Aldrich
TGF-β RI Kinase-Inhibitor IV, CAS-Nr. 909910-43-6, Calbiochem, TGF-β RI Kinase Inhibitor IV, CAS 909910-43-6, is a cell-permeable, selective inhibitor of ALK-4/5/7-mediated signaling (IC50 = 45, 12, and 7.5 nM, respectively).
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat