- On the enzymatic hydrolysis of the sulfate conjugate of 3-methoxy-4-hydroxyphenylglycol (MHPG).
On the enzymatic hydrolysis of the sulfate conjugate of 3-methoxy-4-hydroxyphenylglycol (MHPG).
Biochemical medicine (1980-12-01)
F Karoum, L W Chuang, R J Wyatt
PMID7283987