Merck
  • The Fibronectin-ILT3 Interaction Functions as a Stromal Checkpoint that Suppresses Myeloid Cells.
The Fibronectin-ILT3 Interaction Functions as a Stromal Checkpoint that Suppresses Myeloid Cells.
Cancer immunology research (2021-08-25)
Kevin J Paavola, Julie M Roda, Vicky Y Lin, Peirong Chen, Kyle P O'Hollaren, Richard Ventura, Suzanne C Crawley, Betty Li, Hung-I H Chen, Seth Malmersjö, Nikolai A Sharkov, Geoffrey Horner, Wei Guo, Alan K Kutach, Kalyani Mondal, Zhen Zhang, Joshua S Lichtman, Christina Song, Lee B Rivera, Wenhui Liu, Jian Luo, Yan Wang, Mark J Solloway, Bernard B Allan, Avantika Kekatpure, Shelley R Starck, Raj Haldankar, Bin Fan, Chun Chu, Jie Tang, Martina Molgora, Marco Colonna, Daniel D Kaplan, Jer-Yuan Hsu
ZUSAMMENFASSUNG
Suppressive myeloid cells inhibit antitumor immunity by preventing T-cell responses. Immunoglobulin-like transcript 3 (ILT3; also known as LILRB4) is highly expressed on tumor-associated myeloid cells and promotes their suppressive phenotype. However, the ligand that engages ILT3 within the tumor microenvironment and renders tumor-associated myeloid cells suppressive is unknown. Using a screening approach, we identified fibronectin as a functional ligand for ILT3. The interaction of fibronectin with ILT3 polarized myeloid cells toward a suppressive state, and these effects were reversed with an ILT3-specific antibody that blocked the interaction of ILT3 with fibronectin. Furthermore, ex vivo treatment of human tumor explants with anti-ILT3 reprogrammed tumor-associated myeloid cells toward a stimulatory phenotype. Thus, the ILT3-fibronectin interaction represents a "stromal checkpoint" through which the extracellular matrix actively suppresses myeloid cells. By blocking this interaction, tumor-associated myeloid cells may acquire a stimulatory phenotype, potentially resulting in increased antitumor T-cell responses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polybren Infektions- / Transfektionsreagenz, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Humanplasma Fibronectin-aufgereinigtes Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Humane Hepatische Stellat-LX-2-Zelllinie, a highly suitable model of human hepatic fibrosis
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, ≥98% (HPLC), γ-irradiated, suitable for cell culture