Merck
  • Binding of [125I]Tyr-corticotropin-releasing factor to rabbit aorta is reduced by removal of the endothelium.
Binding of [125I]Tyr-corticotropin-releasing factor to rabbit aorta is reduced by removal of the endothelium.
European journal of pharmacology (1987-03-03)
M R Dashwood, H E Andrews, E T Wei