Merck
  • Single-Cell Transcriptomic Atlas of Different Human Cardiac Arteries Identifies Cell Types Associated With Vascular Physiology.
Single-Cell Transcriptomic Atlas of Different Human Cardiac Arteries Identifies Cell Types Associated With Vascular Physiology.
Arteriosclerosis, thrombosis, and vascular biology (2021-02-26)
Zhan Hu, Wendao Liu, Xiumeng Hua, Xiao Chen, Yuan Chang, Yiqing Hu, Zhenyu Xu, Jiangping Song
ZUSAMMENFASSUNG
[Figure: see text].
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-Aktin-Antikörper , α-Glattmuskel, clone 1A4, ascites fluid