Merck
  • Cholangiopathy and Biliary Fibrosis in Cyp2c70-Deficient Mice Are Fully Reversed by Ursodeoxycholic Acid.
Cholangiopathy and Biliary Fibrosis in Cyp2c70-Deficient Mice Are Fully Reversed by Ursodeoxycholic Acid.
Cellular and molecular gastroenterology and hepatology (2020-12-15)
Jan Freark de Boer, Hilde D de Vries, Anna Palmiotti, Rumei Li, Marwah Doestzada, Joanne A Hoogerland, Jingyuan Fu, Anouk M La Rose, Marit Westerterp, Niels L Mulder, Milaine V Hovingh, Martijn Koehorst, Niels J Kloosterhuis, Justina C Wolters, Vincent W Bloks, Joel T Haas, David Dombrowicz, Bart Staels, Bart van de Sluis, Folkert Kuipers
ZUSAMMENFASSUNG
Bile acids (BAs) aid intestinal fat absorption and exert systemic actions by receptor-mediated signaling. BA receptors have been identified as drug targets for liver diseases. Yet, differences in BA metabolism between humans and mice hamper translation of pre-clinical outcomes. Cyp2c70-ablation in mice prevents synthesis of mouse/rat-specific muricholic acids (MCAs), but potential (patho)physiological consequences of their absence are unknown. We therefore assessed age- and gender-dependent effects of Cyp2c70-deficiency in mice. The consequences of Cyp2c70-deficiency were assessed in male and female mice at different ages. Cyp2c70-/- mice were devoid of MCAs and showed high abundances of chenodeoxycholic and lithocholic acids. Cyp2c70-deficiency profoundly impacted microbiome composition. Bile flow and biliary BA secretion were normal in Cyp2c70-/- mice of both sexes. Yet, the pathophysiological consequences of Cyp2c70-deficiency differed considerably between sexes. Three-week old male Cyp2c70-/- mice showed high plasma BAs and transaminases, which spontaneously decreased thereafter to near-normal levels. Only mild ductular reactions were observed in male Cyp2c70-/- mice up to 8 months of age. In female Cyp2c70-/- mice, plasma BAs and transaminases remained substantially elevated with age, gut barrier function was impaired and bridging fibrosis was observed at advanced age. Addition of 0.1% ursodeoxycholic acid to the diet fully normalized hepatic and intestinal functions in female Cyp2c70-/- mice. Cyp2c70-/- mice show transient neonatal cholestasis and develop cholangiopathic features that progress to bridging fibrosis in females only. These consequences of Cyp2c70-deficiency are restored by treatment with UDCA, indicating a role of BA hydrophobicity in disease development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GAPDH-Maus-mAb (6C5), liquid, clone 6C5, Calbiochem®