Merck
  • Development of HDAC Inhibitors Exhibiting Therapeutic Potential in T-Cell Prolymphocytic Leukemia.
Development of HDAC Inhibitors Exhibiting Therapeutic Potential in T-Cell Prolymphocytic Leukemia.
Journal of medicinal chemistry (2021-06-09)
Krimo Toutah, Nabanita Nawar, Sanna Timonen, Helena Sorger, Yasir S Raouf, Shazreh Bukhari, Jana von Jan, Aleksandr Ianevski, Justyna M Gawel, Olasunkanmi O Olaoye, Mulu Geletu, Ayah Abdeldayem, Johan Israelian, Tudor B Radu, Abootaleb Sedighi, Muzaffar N Bhatti, Muhammad Murtaza Hassan, Pimyupa Manaswiyoungkul, Andrew E Shouksmith, Heidi A Neubauer, Elvin D de Araujo, Tero Aittokallio, Oliver H Krämer, Richard Moriggl, Satu Mustjoki, Marco Herling, Patrick T Gunning
ZUSAMMENFASSUNG
Epigenetic targeting has emerged as an efficacious therapy for hematological cancers. The rare and incurable T-cell prolymphocytic leukemia (T-PLL) is known for its aggressive clinical course. Current epigenetic agents such as histone deacetylase (HDAC) inhibitors are increasingly used for targeted therapy. Through a structure-activity relationship (SAR) study, we developed an HDAC6 inhibitor KT-531, which exhibited higher potency in T-PLL compared to other hematological cancers. KT-531 displayed strong HDAC6 inhibitory potency and selectivity, on-target biological activity, and a safe therapeutic window in nontransformed cell lines. In primary T-PLL patient cells, where HDAC6 was found to be overexpressed, KT-531 exhibited strong biological responses, and safety in healthy donor samples. Notably, combination studies in T-PLL patient samples demonstrated KT-531 synergizes with approved cancer drugs, bendamustine, idasanutlin, and venetoclax. Our work suggests HDAC inhibition in T-PLL could afford sufficient therapeutic windows to achieve durable remission either as stand-alone or in combination with targeted drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Acetyl-Alpha-Tubulin-Antikörper, Klon 6-11B-1, clone 6-11B-1, from mouse
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys18)-Antikörper, serum, Upstate®