Merck
  • Long-term SARS-CoV-2-specific immune and inflammatory responses in individuals recovering from COVID-19 with and without post-acute symptoms.
Long-term SARS-CoV-2-specific immune and inflammatory responses in individuals recovering from COVID-19 with and without post-acute symptoms.
Cell reports (2021-08-07)
Michael J Peluso, Amelia N Deitchman, Leonel Torres, Nikita S Iyer, Sadie E Munter, Christopher C Nixon, Joanna Donatelli, Cassandra Thanh, Saki Takahashi, Jill Hakim, Keirstinne Turcios, Owen Janson, Rebecca Hoh, Viva Tai, Yanel Hernandez, Emily A Fehrman, Matthew A Spinelli, Monica Gandhi, Lan Trinh, Terri Wrin, Christos J Petropoulos, Francesca T Aweeka, Isabel Rodriguez-Barraquer, J Daniel Kelly, Jeffrey N Martin, Steven G Deeks, Bryan Greenhouse, Rachel L Rutishauser, Timothy J Henrich
ZUSAMMENFASSUNG
We describe severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)-specific T cell responses, soluble markers of inflammation, and antibody levels and neutralization capacity longitudinally in 70 individuals with PCR-confirmed SARS-CoV-2 infection. Participants represent a spectrum of illness and recovery, including some with persistent viral shedding in saliva and many experiencing post-acute sequelae of SARS-CoV-2 infection (PASC). T cell responses remain stable for up to 9 months. Whereas the magnitude of early CD4+ T cell immune responses correlates with severity of initial infection, pre-existing lung disease is independently associated with higher long-term SARS-CoV-2-specific CD8+ T cell responses. Among participants with PASC 4 months following coronavirus disease 2019 (COVID-19) symptom onset, we observe a lower frequency of CD8+ T cells expressing CD107a, a marker of degranulation, in response to Nucleocapsid (N) peptide pool stimulation, and a more rapid decline in the frequency of N-specific interferon-γ-producing CD8+ T cells. Neutralizing antibody levels strongly correlate with SARS-CoV-2-specific CD4+ T cell responses.