Merck
Variability in the Munc13-1 content of excitatory release sites.
eLife (2021-04-28)
Maria Rita Karlocai, Judit Heredi, Tünde Benedek, Noemi Holderith, Andrea Lorincz, Zoltan Nusser
ZUSAMMENFASSUNG
The molecular mechanisms underlying the diversity of cortical glutamatergic synapses are still incompletely understood. Here, we tested the hypothesis that presynaptic active zones (AZs) are constructed from molecularly uniform, independent release sites (RSs), the number of which scales linearly with the AZ size. Paired recordings between hippocampal CA1 pyramidal cells and fast-spiking interneurons in acute slices from adult mice followed by quantal analysis demonstrate large variability in the number of RSs (N) at these connections. High-resolution molecular analysis of functionally characterized synapses reveals variability in the content of one of the key vesicle priming factors - Munc13-1 - in AZs that possess the same N. Replica immunolabeling also shows a threefold variability in the total Munc13-1 content of AZs of identical size and a fourfold variability in the size and density of Munc13-1 clusters within the AZs. Our results provide evidence for quantitative molecular heterogeneity of RSs and support a model in which the AZ is built up from variable numbers of molecularly heterogeneous, but independent RSs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Albumin aus Rinderserum, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Trizma® Hydrochlorid, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
Adenosin-5′-triphosphat Dinatriumsalz Hydrat, Grade I, ≥99%, from microbial
Sigma-Aldrich
Guanosin 5′-triphosphat Natriumsalz Hydrat, ≥95% (HPLC), powder
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Magnesiumchlorid Hexahydrat, BioXtra, ≥99.0%
Sigma-Aldrich
Calciumchlorid Dihydrat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumkreatinphosphat, zweibasisch Dinatriumsalz Tetrahydrat, ≥98.0% (NT)