Merck
  • An optimized protocol for patient-derived xenograft in humanized mice to evaluate the role of IL-34 in immunotherapeutic resistance.
An optimized protocol for patient-derived xenograft in humanized mice to evaluate the role of IL-34 in immunotherapeutic resistance.
STAR protocols (2021-04-27)
Nanumi Han, Hye Yoon Jang, Naoki Hama, Takuto Kobayashi, Ryo Otsuka, Haruka Wada, Ken-Ichiro Seino
ZUSAMMENFASSUNG
Previously, we identified a therapy-resistant role of IL-34 in an immune checkpoint blockade in murine models. To investigate whether a similar mechanism is applicable in human tumors as well, we used this protocol for the selection of IL-34-neutralizing antibody and transplanting human tumor tissue expressing both IL-34 and PD-L1 as a patient-derived xenograft in immunologically humanized mice. This model helps to determine the effect of IL-34 neutralization along with the immune checkpoint blockade in human tumors. For complete details on the use and execution of this protocol, please refer to Hama et al. (2020).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Fetales Kälberserum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-IL-34, clone 1D12 Antibody, ascites fluid, clone 1D12, from mouse