Merck
Inducible ablation of CD11c+ cells to determine their role in skin wound repair.
Immunology (2021-01-28)
Zhi Li, Rebecca Lamb, Mark C Coles, Clare L Bennett, Carrie A Ambler
ZUSAMMENFASSUNG
Whether resident and recruited myeloid cells may impair or aid healing of acute skin wounds remains a debated question. To begin to address this, we examined the importance of CD11c+ myeloid cells in the early activation of skin wound repair. We find that an absence of CD11c+ cells delays wound closure and epidermal proliferation, likely due to defects in the activation of the IL-23-IL-22 axis that is required for wound healing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DNAse I, grade II, from bovine pancreas
Sigma-Aldrich
Diphterietoxin aus Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein