Merck
  • Chemotactic TEG3 Cells' Guiding Platforms Based on PLA Fibers Functionalized With the SDF-1α/CXCL12 Chemokine for Neural Regeneration Therapy.
Chemotactic TEG3 Cells' Guiding Platforms Based on PLA Fibers Functionalized With the SDF-1α/CXCL12 Chemokine for Neural Regeneration Therapy.
Frontiers in bioengineering and biotechnology (2021-04-09)
Oscar Castaño, Ana López-Mengual, Diego Reginensi, Andreu Matamoros-Angles, Elisabeth Engel, José Antonio Del Rio
ZUSAMMENFASSUNG
(Following spinal cord injury, olfactory ensheathing cell (OEC) transplantation is a promising therapeutic approach in promoting functional improvement. Some studies report that the migratory properties of OECs are compromised by inhibitory molecules and potentiated by chemical concentration differences. Here we compare the attachment, morphology, and directionality of an OEC-derived cell line, TEG3 cells, seeded on functionalized nanoscale meshes of Poly(l/dl-lactic acid; PLA) nanofibers. The size of the nanofibers has a strong effect on TEG3 cell adhesion and migration, with the PLA nanofibers having a 950 nm diameter being the ones that show the best results. TEG3 cells are capable of adopting a bipolar morphology on 950 nm fiber surfaces, as well as a highly dynamic behavior in migratory terms. Finally, we observe that functionalized nanofibers, with a chemical concentration increment of SDF-1α/CXCL12, strongly enhance the migratory characteristics of TEG3 cells over inhibitory substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laminin aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
Phalloidin-Fluoresceinisothiocyanat-Konjugat aus Amanita phalloides, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
bisBenzimid H 33342, ≥98% (HPLC)
Sigma-Aldrich
Anti-Integrin α5β1 Antibody, clone BMB5, ascites fluid, clone BMB5, Chemicon®