Merck
  • Determination of very-long-chain fatty acids in plasma by a simplified gas chromatographic-mass spectrometric procedure.
Determination of very-long-chain fatty acids in plasma by a simplified gas chromatographic-mass spectrometric procedure.
Journal of Chromatography A (1991)
Caruso U, Fowler B, Erceg M, Romano C.