Merck
Chromosomal instability by mutations in the novel minor spliceosome component CENATAC.
The EMBO journal (2021-05-20)
Bas de Wolf, Ali Oghabian, Maureen V Akinyi, Sandra Hanks, Eelco C Tromer, Jolien J E van Hooff, Lisa van Voorthuijsen, Laura E van Rooijen, Jens Verbeeren, Esther C H Uijttewaal, Marijke P A Baltissen, Shawn Yost, Philippe Piloquet, Michiel Vermeulen, Berend Snel, Bertrand Isidor, Nazneen Rahman, Mikko J Frilander, Geert J P L Kops
ZUSAMMENFASSUNG
Aneuploidy is the leading cause of miscarriage and congenital birth defects, and a hallmark of cancer. Despite this strong association with human disease, the genetic causes of aneuploidy remain largely unknown. Through exome sequencing of patients with constitutional mosaic aneuploidy, we identified biallelic truncating mutations in CENATAC (CCDC84). We show that CENATAC is a novel component of the minor (U12-dependent) spliceosome that promotes splicing of a specific, rare minor intron subtype. This subtype is characterized by AT-AN splice sites and relatively high basal levels of intron retention. CENATAC depletion or expression of disease mutants resulted in excessive retention of AT-AN minor introns in ˜ 100 genes enriched for nucleocytoplasmic transport and cell cycle regulators, and caused chromosome segregation errors. Our findings reveal selectivity in minor intron splicing and suggest a link between minor spliceosome defects and constitutional aneuploidy in humans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-CCDC84 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody