Merck
  • Induction of mast cell proliferation, maturation, and heparin synthesis by the rat c-kit ligand, stem cell factor
Induction of mast cell proliferation, maturation, and heparin synthesis by the rat c-kit ligand, stem cell factor
Proceedings of the National Academy of Sciences of the USA (1991)
Tsai M et al