Merck
  • Retinal Proteomics of a Mouse Model of Dystroglycanopathies Reveals Molecular Alterations in Photoreceptors.
Retinal Proteomics of a Mouse Model of Dystroglycanopathies Reveals Molecular Alterations in Photoreceptors.
Journal of proteome research (2021-05-25)
Mary Luz Uribe, José Martín-Nieto, Cristina Quereda, Marcos Rubio-Fernández, Jesús Cruces, George M C Janssen, Arnoud H de Ru, Peter A van Veelen, Paul J Hensbergen
ZUSAMMENFASSUNG
Mutations in the POMT1 gene, encoding a protein O-mannosyltransferase essential for α-dystroglycan (α-DG) glycosylation, are frequently observed in a group of rare congenital muscular dystrophies, collectively known as dystroglycanopathies. However, it is hitherto unclear whether the effects seen in affected patients can be fully ascribed to α-DG hypoglycosylation. To study this, here we used comparative mass spectrometry-based proteomics and immunofluorescence microscopy and investigated the changes in the retina of mice in which Pomt1 is specifically knocked out in photoreceptor cells. Our results demonstrate significant proteomic changes and associated structural alteration in photoreceptor cells of Pomt1 cKO mice. In addition to the effects related to impaired α-DG O-mannosylation, we observed morphological alterations in the outer segment that are associated with dysregulation of a relatively understudied POMT1 substrate (KIAA1549), BBSome proteins, and retinal stress markers. In conclusion, our study provides new hypotheses to explain the phenotypic changes that are observed in the retina of patients with dystroglycanopathies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-GFAP-Antikörper (Glial Fibrillary Acidic Protein, Saures Gliafaserprotein) in Maus hergestellte Antikörper, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Arrestin-C-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Rhodopsin Antibody, CT, last 9 amino acids, clone Rho 1D4, clone Rho 1D4, Chemicon®, from mouse
Sigma-Aldrich
Anti-FGF-2/basic-FGF-Antikörper, Klon bFM-2, clone bFM-2, Upstate®, from mouse