Merck
  • Effect of human transferrin on the engraftment of allogeneic bone marrow in various strans of lethally irradiated mice.
Effect of human transferrin on the engraftment of allogeneic bone marrow in various strans of lethally irradiated mice.
Nature Immunology (1992)
Pierpaoli W.