Merck
Expression cloning of the murine and human interleukin 9 receptor cDNAs.
Proceedings of the National Academy of Sciences of the United States of America (1992-06-15)
J C Renauld, C Druez, A Kermouni, F Houssiau, C Uyttenhove, E Van Roost, J Van Snick
ZUSAMMENFASSUNG
Interleukin 9 (IL-9) is a T-cell-derived lymphokine that induces the proliferation of various lymphoid and hemopoietic cells. A cDNA clone encoding the murine IL-9 receptor was isolated by expression cloning in COS cells and screening with 125I-labeled IL-9. Transient expression of this cDNA produced high-affinity binding sites for IL-9. The predicted 52-kDa protein contains a putative signal peptide and a typical transmembrane domain. A cDNA for the human homologue was isolated by cross-hybridization. Transfection of this cDNA in a murine T-cell clone conferred responsiveness to human IL-9. Sequence analysis revealed that the IL-9 receptor belongs to the recently described hematopoietin receptor super-family and is expressed in membrane-bound and soluble forms.