Merck
  • Blood-based protein profiling identifies serum protein c-KIT as a novel biomarker for hypertrophic cardiomyopathy.
Blood-based protein profiling identifies serum protein c-KIT as a novel biomarker for hypertrophic cardiomyopathy.
Scientific reports (2021-01-21)
Kristina Sonnenschein, Jan Fiedler, David de Gonzalo-Calvo, Ke Xiao, Angelika Pfanne, Annette Just, Carolin Zwadlo, Samira Soltani, Udo Bavendiek, Theresia Kraft, Cristobal Dos Remedios, Serghei Cebotari, Johann Bauersachs, Thomas Thum
ZUSAMMENFASSUNG
Hypertrophic cardiomyopathy (HCM) is one of the most common hereditary heart diseases and can be classified into an obstructive (HOCM) and non-obstructive (HNCM) form. Major characteristics for HCM are the hypertrophy of cardiomyocytes and development of cardiac fibrosis. Patients with HCM have a higher risk for sudden cardiac death compared to a healthy population. In the present study, we investigated the abundancy of selected proteins as potential biomarkers in patients with HCM. We included 60 patients with HCM and 28 healthy controls and quantitatively measured the rate of a set of 92 proteins already known to be associated with cardiometabolic processes via protein screening using the proximity extension assay technology in a subgroup of these patients (20 HCM and 10 healthy controls). After validation of four hits in the whole cohort of patients consisting of 88 individuals (60 HCM patients, 28 healthy controls) we found only one candidate, c-KIT, which was regulated significantly different between HCM patients and healthy controls and thus was chosen for further analyses. c-KIT is a tyrosine-protein kinase acting as receptor for the stem cell factor and activating several pathways essential for cell proliferation and survival, hematopoiesis, gametogenesis and melanogenesis. Serum protein levels of c-KIT were significantly lower in patients with HCM than in healthy controls, even after adjusting for confounding factors age and sex. In addition, c-KIT levels in human cardiac tissue of patients with HOCM were significant higher compared to controls indicating high levels of c-KIT in fibrotic myocardium. Furthermore, c-KIT concentration in serum significantly correlated with left ventricular end-diastolic diameter in HOCM, but not HCM patients. The present data suggest c-KIT as a novel biomarker differentiating between patients with HCM and healthy population and might provide further functional insights into fibrosis-related processes of HOCM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Human KIT / Mast/Stem Cell Growth Factor Receptor Kit ELISA Kit
Sigma-Aldrich
Human PCPE-1 ELISA, for serum, plasma and cell culture supernatants