Merck
  • Premigratory neural crest stem cells generate enteric neurons populating the mouse colon and regulating peristalsis in tissue-engineered intestine.
Premigratory neural crest stem cells generate enteric neurons populating the mouse colon and regulating peristalsis in tissue-engineered intestine.
Stem cells translational medicine (2021-01-23)
Huipu Yuan, Hui Hu, Rui Chen, Wenbo Mu, Liangliang Wang, Ying Li, Yuelei Chen, Xiaoyan Ding, Yongmei Xi, ShanShan Mao, Mizu Jiang, Jie Chen, Yong He, Lang Wang, Yi Dong, Jinfa Tou, Wei Chen
ZUSAMMENFASSUNG
Hirschsprung's disease (HSCR) is a common congenital defect. It occurs when bowel colonization by neural crest-derived enteric nervous system (ENS) precursors is incomplete during the first trimester of pregnancy. Several sources of candidate cells have been previously studied for their capacity to regenerate the ENS, including enteric neural crest stem cells (En-NCSCs) derived from native intestine or those simulated from human pluripotent stem cells (hPSCs). However, it is not yet known whether the native NCSCs other than En-NCSCs would have the potential of regenerating functional enteric neurons and producing neuron dependent motility under the intestinal environment. The present study was designed to determine whether premigratory NCSCs (pNCSCs), as a type of the nonenteric NCSCs, could form enteric neurons and mediate the motility. pNCSCs were firstly transplanted into the colon of adult mice, and were found to survive, migrate, differentiate into enteric neurons, and successfully integrate into the adult mouse colon. When the mixture of pNCSCs and human intestinal organoids was implanted into the subrenal capsule of nude mice and grown into the mature tissue-engineered intestine (TEI), the pNCSCs-derived neurons mediated neuron-dependent peristalsis of TEI. These results show that the pNCSCs that were previously assumed to not be induced by intestinal environment or cues can innervate the intestine and establish neuron-dependent motility. Future cell candidates for ENS regeneration may include nonenteric NCSCs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cholin-Acetyltransferase-Antikörper, Chemicon®, from goat
Sigma-Aldrich
Anti-Neurofilament 200 in Kaninchen hergestellte Antikörper, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Nestin-Antikörper, Klon 401 (Ratte), clone rat-401, Chemicon®, from mouse